missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC434091000

 View more versions of this product

Catalog No. AC434091000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
96.0% min.
White to Yellow
100mg
96.0
97%
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)N
381.459
381.45
97%
Authentic
Glass bottle
87.0°C to 93.0°C
1091606-67-5
100.0
C26H24NP
MIPGTOGPLXZBQX-UIOOFZCWSA-N
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
46177587
Crystalline Powder

Chemical Identifiers

1091606-67-5
381.459
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
C26H24NP
MIPGTOGPLXZBQX-UIOOFZCWSA-N
46177587
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)N
SDS Documents