Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-(+)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 99%

$223.00 - $223.00

Chemical Identifiers

Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentylamine
CAS 181657-57-8
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD01075751
UN Number UN2735
Synonym (1S,2S)-1-Amino-2-benzyloxycyclopentane; (1S-trans)-2-(Phenylmethoxy)cyclopentanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1701806

  SDS
alfa aesar™
L1701806
5g Each for $223.00

AAL1701803

  SDS
alfa aesar™
L1701803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.27
Assay Percent Notes ee 99%
Molecular Formula C12H17NO
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 90° to 93°C (0.23mmHg)
Quantity 5g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Assay ≥99%
MDL Number MFCD01075751
UN Number UN2735
Synonym (1S,2S)-1-Amino-2-benzyloxycyclopentane; (1S-trans)-2-(Phenylmethoxy)cyclopentanamine
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentylamine
Optical Rotation +57.5° (Neat)
Health Hazard 1 Danger
CAS 181657-57-8
Melting Point 23°C
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P309-P310-P321
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).

Applications
(1S,2S)-(+)-2-Benzyloxycyclopentylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air and carbon dioxide.

Safety and Handling

Danger

GHS P Statement
P280-P264-P305+P351+P338-P309-P310-P321
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.