Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-(+)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 99%


Quantity 5g

Manufacturer: alfa aesar™  L1701806

 View more versions of this product

Catalog No. AAL1701806

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 191.27
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentylamine
Assay ≥99%
Assay Percent Notes ee 99%
Molecular Formula C12H17NO
MDL Number MFCD01075751
CAS 181657-57-8
UN Number UN2735
Synonym (1S,2S)-1-Amino-2-benzyloxycyclopentane; (1S-trans)-2-(Phenylmethoxy)cyclopentanamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 90° to 93°C (0.23mmHg)
Quantity 5g
Optical Rotation +57.5° (Neat)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P309-P310-P321
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Melting Point 23°C
Flash Point >110°C (230°F)

Specifications

Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentylamine
CAS 181657-57-8
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD01075751
UN Number UN2735
Synonym (1S,2S)-1-Amino-2-benzyloxycyclopentane; (1S-trans)-2-(Phenylmethoxy)cyclopentanamine

Applications
(1S,2S)-(+)-2-Benzyloxycyclopentylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air and carbon dioxide.

Accessories

Danger

GHS P Statement
P280-P264-P305+P351+P338-P309-P310-P321
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.