Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-(+)-2-Benzyloxycyclopentyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentyl isothiocyanate
CAS 737000-91-8
Molecular Formula C13H15NOS
Formula Weight 233.33
MDL Number MFCD05664049
UN Number UN2922
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2006203

  SDS
alfa aesar™
L2006203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.33
MDL Number MFCD05664049
Health Hazard 1 Danger
Molecular Formula C13H15NOS
Assay 97%
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
UN Number UN2922
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Recommended Storage 2° to 8°C
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentyl isothiocyanate
CAS 737000-91-8
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

2° to 8°C