Learn More
(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1702006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(1S,2S)-(+)-2-Benzyloxycyclohexylamine | |
1.02 | |
>110°C (230°F) | |
C13H19NO | |
MFCD01075753 | |
UN2735 | |
Air Sensitive | |
NTHNRYLIXJZHRZ-UHFFFAOYNA-N | |
NC1CCCCC1OCC1=CC=CC=C1 | |
205.30 | |
205.3 |
216394-07-9 | |
79°C to 80°C (0.08 mmHg) | |
ee 99% | |
1.5275 | |
5 g | |
8981794 | |
1s,2s-2-benzyloxy cyclohexanamine, 1s,2s-+-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxy cyclohexan-1-amine, 1s,2s-trans-2-benzyloxy-cyclohexylamine, 1s, 2s-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxy-cyclohexylamine, 1s, 2s-2-benzyloxy-cyclohexylamine, 1s,2s-2-benzyloxy cyclohexylamine, 1s,2s-1-amino-2-benzyloxycyclohexane | |
+85° (Neat) | |
(1S,2S)-2-phenylmethoxycyclohexan-1-amine | |
11908838 | |
≥99% |
Chemical Identifiers
216394-07-9 | |
205.30 | |
NTHNRYLIXJZHRZ-UHFFFAOYNA-N | |
(1S,2S)-2-phenylmethoxycyclohexan-1-amine |
C13H19NO | |
MFCD01075753 | |
11908838 | |
NC1CCCCC1OCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 606-811-1
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only