Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-(+)-2-Benzyloxycyclohexyl isocyanate, 97%

$150.68 - $150.68

Chemical Identifiers

Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclohexyl isocyanate
CAS 361392-21-4
Molecular Formula C14H17NO2
Formula Weight 231.3
MDL Number MFCD05664068
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2022303

  SDS
alfa aesar™
L2022303
1g Each for $150.68
Description & Specifications

Specifications

Formula Weight 231.3
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclohexyl isocyanate
Assay 97%
Molecular Formula C14H17NO2
MDL Number MFCD05664068
CAS 361392-21-4
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C