Please login to your online account to display your discounted pricing

(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  181490050

 View more versions of this product

Catalog No. AC181490050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Formula Weight 167.21
Assay ≥97.5%
Quantity 5g
Color White to yellow
TSCA TSCA
EINECS Number 248-867-6
Melting Point 110° to 114°C
CAS Max % 100.0
Packaging Glass bottle
Specific Rotation +35° to +39° (20°C, 589nm) (c=1, 1N HCl)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Chemical Name or Material (1S, 2S)-(+)-2-Amino-1-phenyl-1, 3-propanediol
CAS 28143-91-1
CAS Min % 97.5
Name Note 98%
Molecular Formula C9H13NO2
MDL Number MFCD00004503
Linear Formula C6H5CH(OH)CH(NH2)CH2OH
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material (1S, 2S)-(+)-2-Amino-1-phenyl-1, 3-propanediol
Name Note 98%
CAS 28143-91-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13NO2
Linear Formula C6H5CH(OH)CH(NH2)CH2OH
Formula Weight 167.21
MDL Number MFCD00004503
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents