Please login to your online account to display your discounted pricing

(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334800250

 View more versions of this product

Catalog No. AC334800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 212.2
Quantity 25g
Color Yellow to beige
Melting Point 160° to 166°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C9H12N2O4
Packaging Glass bottle
Chemical Name or Material (1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
CAS 2964-48-9
Specific Rotation +29° to +33° (20°C, 589nm) (c=1, 6N HCl)
MDL Number MFCD00007359
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
CAS 2964-48-9
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H12N2O4
Formula Weight 212.2
MDL Number MFCD00007359