missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2S)-1-Amino-2-(diphenylphosphino)indane, 97+%

$61.05 - $272.96

Chemical Identifiers

CAS 1091606-69-7
Molecular Formula C21H20NP
Molecular Weight (g/mol) 317.372
MDL Number MFCD17013984
InChI Key IALYUOSURKZWLR-SFTDATJTSA-N
Synonym 1s,2s-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine, 1s,2s-1-amino-2-diphenylphosphino indane, 1s,2s-2-diphenylphosphanyl-2,3-dihydro-1h-inden-1-amine, 1s,2s-2-diphenylphosphino indan-1-amine, 1s,2s-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine,min 10wt% in thf
PubChem CID 46177588
IUPAC Name (1S,2S)-2-diphenylphosphanyl-2,3-dihydro-1H-inden-1-amine
SMILES C1C(C(C2=CC=CC=C21)N)P(C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6067103 View Documents Alfa Aesar™
H6067103
1g Each for $61.05
 
AAH6067106 View Documents Alfa Aesar™
H6067106
5g Each for $272.96
 
Specifications

Chemical Identifiers

1091606-69-7
317.372
IALYUOSURKZWLR-SFTDATJTSA-N
46177588
C1C(C(C2=CC=CC=C21)N)P(C3=CC=CC=C3)C4=CC=CC=C4
C21H20NP
MFCD17013984
1s,2s-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine, 1s,2s-1-amino-2-diphenylphosphino indane, 1s,2s-2-diphenylphosphanyl-2,3-dihydro-1h-inden-1-amine, 1s,2s-2-diphenylphosphino indan-1-amine, 1s,2s-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine,min 10wt% in thf
(1S,2S)-2-diphenylphosphanyl-2,3-dihydro-1H-inden-1-amine

Specifications

1g
C21H20NP
1s,2s-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine, 1s,2s-1-amino-2-diphenylphosphino indane, 1s,2s-2-diphenylphosphanyl-2,3-dihydro-1h-inden-1-amine, 1s,2s-2-diphenylphosphino indan-1-amine, 1s,2s-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine,min 10wt% in thf
C1C(C(C2=CC=CC=C21)N)P(C3=CC=CC=C3)C4=CC=CC=C4
317.372
317.36
(1S,2S)-1-Amino-2-(diphenylphosphino)indane
1091606-69-7
MFCD17013984
IALYUOSURKZWLR-SFTDATJTSA-N
(1S,2S)-2-diphenylphosphanyl-2,3-dihydro-1H-inden-1-amine
46177588
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning