missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine 97.0+%, TCI America™
$192.44 - $575.67
Chemical Identifiers
CAS | 186769-18-6 |
---|---|
Molecular Formula | C20H28N2 |
Molecular Weight (g/mol) | 296.458 |
MDL Number | MFCD06797064 |
InChI Key | ILMRHFMYIXTNMC-PMACEKPBSA-N |
Synonym | (1S,2S)-1,2-Diamino-1,2-dimesitylethane, (1S,2S)-1,2-Dimesitylethylenediamine |
PubChem CID | 11289465 |
IUPAC Name | (1S,2S)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine |
SMILES | CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B2317100MG
|
TCI America
B2317100MG |
100 mg |
Each for $192.44
|
|
|||||
B2317500MG
|
TCI America
B2317500MG |
500 mg |
Each for $575.67
|
|
|||||
Chemical Identifiers
186769-18-6 | |
296.458 | |
ILMRHFMYIXTNMC-PMACEKPBSA-N | |
11289465 | |
CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C |
C20H28N2 | |
MFCD06797064 | |
(1S,2S)-1,2-Diamino-1,2-dimesitylethane, (1S,2S)-1,2-Dimesitylethylenediamine | |
(1S,2S)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine |
Specifications
186769-18-6 | |
White-Yellow | |
MFCD06797064 | |
(1S,2S)-1,2-Diamino-1,2-dimesitylethane, (1S,2S)-1,2-Dimesitylethylenediamine | |
CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C | |
296.458 | |
296.46 | |
Crystalline Powder |
85°C | |
C20H28N2 | |
100 mg | |
ILMRHFMYIXTNMC-PMACEKPBSA-N | |
(1S,2S)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine | |
11289465 | |
≥97.0% (GC,T) | |
(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine |
Safety and Handling
TSCA : No