missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine, TCI America™

$348.34 - $1,194.75

Chemical Identifiers

CAS 365996-30-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD09952105
InChI Key AKVIZYGPJIWKOS-BDAKNGLRSA-N
Synonym (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
PubChem CID 1514389
IUPAC Name tert-butyl N-[(1S,2R)-2-aminocyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCCC1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34831G View Documents TCI America
B34831G
1g Each for $348.34
 
B34835G View Documents TCI America
B34835G
5g Each for $1,194.75
 
Specifications

Chemical Identifiers

365996-30-1
214.309
AKVIZYGPJIWKOS-BDAKNGLRSA-N
1514389
CC(C)(C)OC(=O)NC1CCCCC1N
C11H22N2O2
MFCD09952105
(1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
tert-butyl N-[(1S,2R)-2-aminocyclohexyl]carbamate

Specifications

White
365996-30-1
MFCD09952105
AKVIZYGPJIWKOS-BDAKNGLRSA-N
tert-butyl N-[(1S,2R)-2-aminocyclohexyl]carbamate
1514389
Crystal-Lump at 20°C
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
1g
C11H22N2O2
(1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
CC(C)(C)OC(=O)NC1CCCCC1N
214.309
214.31
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No