Please login to your online account to display your discounted pricing

(1S,2R,4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1S, 2R, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
CAS 321744-23-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD02259719
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC362305000

  SDS
fisher scientific acros
362305000
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Moist Crystals
Formula Weight 259.3
Quantity 500mg
Color Off-white to yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material (1S, 2R, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C12H21NO5
MDL Number MFCD02259719
CAS 321744-23-4
Packaging Glass bottle
Synonym Methyl (1R, 3S, 4R)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate
Enantiomeric Excess 97.5% min. (GC)