Please login to your online account to display your discounted pricing

(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride
Name Note 95%, (98% e.e.)
CAS 220497-88-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H13NO3·HCl
Formula Weight 183.63
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362530010

  SDS
thermo fisher scientific
AC362530010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride
Color White to off-white
Quantity 1g
Name Note 95%, (98% e.e.)
CAS 220497-88-1
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Enantiomeric Excess 97.5% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
TSCA TSCA
Packaging Glass bottle
Physical Form Crystalline Powder
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H13NO3·HCl
Formula Weight 183.63
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.