Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol

$84.36 - $84.36

Chemical Identifiers

Chemical Name or Material (1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol
CAS 123620-80-4
Molecular Formula C13H19NO
Formula Weight 205.30g/mol
MDL Number MFCD02093492
Synonym (1S,2R)-N-Pyrrolidinylnorephedrine base; (1S,2R)-N,N-Tetramethylenenorephedrine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3124403

 
alfa aesar™
H31244-03
1g Each for $84.36
Description & Specifications

Specifications

Melting Point 44° to 46°C
Boiling Point 155°C (3mmHg)
Quantity 1g
Chemical Name or Material (1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol
CAS 123620-80-4
Molecular Formula C13H19NO
Formula Weight 205.30g/mol
MDL Number MFCD02093492
Synonym (1S,2R)-N-Pyrrolidinylnorephedrine base; (1S,2R)-N,N-Tetramethylenenorephedrine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.