Please login to your online account to display your discounted pricing

Alfa Aesar™ (1S)-10-Camphorsulfonamide, 97%

$184.72 - $788.11

Chemical Identifiers

Chemical Name or Material (1S)-10-Camphorsulfonamide
CAS 60933-63-3
Molecular Formula C10H17NO3S
Formula Weight 231.32
MDL Number MFCD00075611
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4436106

  SDS
alfa aesar™
4436106
5g Each for $184.72

AA4436114

  SDS
alfa aesar™
4436114
25g Each for $788.11
Description & Specifications

Specifications

Formula Weight 231.32
Chemical Name or Material (1S)-10-Camphorsulfonamide
Physical Form Powder
Assay 97%
Molecular Formula C10H17NO3S
MDL Number MFCD00075611
CAS 60933-63-3
Melting Point 133° to 135°C (dec.)
Quantity 5g