missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R)-(-)-Nopol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 129200050

 View more versions of this product

Catalog No. AC129200050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(1R)-(-)-Nopol
− 37.00 (23.00°C neat)
5mL
35836-73-8
100.0
Glass bottle
-36° to -39° (20°C, 589nm) (C=neat)
≥97.5%
C11H18O
MFCD00075187
(1R, 5S)-6, 6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol
0.9730g/mL
Clear colorless to light yellow
98%
97.5
Authentic
1.4920 to 1.4940 (20°C, 589nm)
Liquid
98%
166.26
15, 6771

Chemical Identifiers

35836-73-8
100.0
C11H18O
MFCD00075187
(1R)-(-)-Nopol, 98%
97.5
98%
166.26
(1R, 5S)-6, 6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-890-3

RTECSNumber : RC8870000

TSCA : TSCA