Please login to your online account to display your discounted pricing

(1R)-(-)-Nopol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1R)-(-)-Nopol
Name Note 98%
CAS 35836-73-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H18O
Formula Weight 166.26
MDL Number MFCD00075187
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC129200050

  SDS
thermo fisher scientific
129200050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 166.26
Quantity 5mL
Color Clear colorless to light yellow
EINECS Number 204-890-3
TSCA TSCA
RTECS Number RC8870000
Density 0.9730g/mL
Specific Rotation Condition − 37.00 (23.00°C neat)
Merck Index 15, 6771
CAS Max % 100.0
Chemical Name or Material (1R)-(-)-Nopol
Packaging Glass bottle
Name Note 98%
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Specific Rotation -36° to -39° (20°C, 589nm) (C=neat)
Assay Percent Range 98%
Molecular Formula C11H18O
MDL Number MFCD00075187
CAS 35836-73-8
Synonym (1R, 5S)-6, 6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.