Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R)-(-)-Menthyl chloroformate, 90+%

$56.50 - $56.50

Chemical Identifiers

Chemical Name or Material (1R)-(-)-Menthyl chloroformate
CAS 14602-86-9
Molecular Formula C11H19ClO2
Formula Weight 218.73
MDL Number MFCD00009694
UN Number UN3277
Beilstein 2414686
Synonym Chloroformic acid (1R)-menthyl ester; L-Menthyl chloroformate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1173906

  SDS
alfa aesar™
L1173906*
5g Each for $56.50

AAL1173914

  SDS
alfa aesar™
L1173914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.73
Assay 95%
Molecular Formula C11H19ClO2
MDL Number MFCD00009694
CAS 14602-86-9
UN Number UN3277
Beilstein 2414686
Boiling Point 108° to 109°C (11mmHg)
Synonym Chloroformic acid (1R)-menthyl ester; L-Menthyl chloroformate
Quantity 5g
Flash Point 70°C (158°F)
Optical Rotation −82° (c=1 in Chloroform)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, N.O.S.
Chemical Name or Material (1R)-(-)-Menthyl chloroformate
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

2° to 8°C