missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R)-(-)-Menthyl chloroformate, 90+%

Manufacturer:  Alfa Aesar™ L1173906*

 View more versions of this product

Catalog No. AAL1173906



Specifications

Specifications

(1R)-(-)-Menthyl chloroformate
−82° (c=1 in Chloroform)
5g
C11H19ClO2
UN3277
1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
CC1CCC(C(C1)OC(=O)Cl)C(C)C
218.721
218.73
70°C (158°F)
108°C to 109°C (11mmHg)
14602-86-9
MFCD00009694
2414686
KIUPCUCGVCGPPA-KXUCPTDWSA-N
[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate
7014897
95%

Chemical Identifiers

14602-86-9
218.721
KIUPCUCGVCGPPA-KXUCPTDWSA-N
7014897
CC1CCC(C(C1)OC(=O)Cl)C(C)C
C11H19ClO2
MFCD00009694
1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, N.O.S.

Recommended Storage : 2° to 8°C

Documents