Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R)-(-)-Camphorquinone, 98%

$131.39 - $131.39

Chemical Identifiers

Chemical Name or Material (1R)-(-)-Camphorquinone
CAS 10334-26-6
Molecular Formula C10H14O2
Formula Weight 166.22
MDL Number MFCD00082863
Beilstein 2327696
Synonym (1R)-(-)-2,3-Bornanedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2409206

  SDS
alfa aesar™
B2409206
5g Each for $131.39
Description & Specifications

Specifications

Formula Weight 166.22
Chemical Name or Material (1R)-(-)-Camphorquinone
Assay 98%
Molecular Formula C10H14O2
MDL Number MFCD00082863
CAS 10334-26-6
Beilstein 2327696
Synonym (1R)-(-)-2,3-Bornanedione
Melting Point 197° to 202°C
Quantity 5g
Optical Rotation −102° (c=2 in Ethanol)