Learn More
(1R)-(+)-Camphor, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1070814
Description
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
Solubility
Soluble in water (0.1 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
(1R)- (+)-Camphor | |
175°C to 180°C | |
204°C | |
Camphor-like | |
MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149 | |
UN2717 | |
14,1732 | |
Soluble in water (0.1g/L at 20°C). | |
+43° (c=10 in ethanol) | |
(1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one | |
159055 | |
152.24 |
464-49-3 | |
0.99 | |
65°C (149°F) | |
C10H16O | |
25 g | |
2042745 | |
+-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone | |
DSSYKIVIOFKYAU-XVKPBYJWSA-N | |
CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2 | |
152.24 | |
CHEBI:15396 | |
98% |
Chemical Identifiers
464-49-3 | |
152.24 | |
DSSYKIVIOFKYAU-XVKPBYJWSA-N | |
159055 | |
(1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
C10H16O | |
MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149 | |
+-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone | |
CHEBI:15396 | |
CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2 |
Safety and Handling
GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR
EINECSNumber : 207-355-2
RTECSNumber : EX1260000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only