Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R)-(-)-Camphor-10-sulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material (1R)-(-)-Camphor-10-sulfonyl chloride
CAS 39262-22-1
Molecular Formula C10H15ClO3S
Formula Weight 250.74
MDL Number MFCD00064155
UN Number UN3261
Beilstein 3205975
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1414503

  SDS
alfa aesar™
L1414503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1414506

  SDS
alfa aesar™
L1414506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1414514

  SDS
alfa aesar™
L1414514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.74
Chemical Name or Material (1R)-(-)-Camphor-10-sulfonyl chloride
Assay 97%
Molecular Formula C10H15ClO3S
MDL Number MFCD00064155
CAS 39262-22-1
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN3261
Beilstein 3205975
Melting Point 64° to 68°C
Optical Rotation −32° (c=1 in Chloroform)
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C