Please login to your online account to display your discounted pricing

(1R,4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1R, 4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid
CAS 298716-03-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H17NO4
Formula Weight 227.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC435880010

  SDS
thermo fisher scientific
AC435880010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.26
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 1g
Color White
Melting Point 110° to 115°C
CAS Max % 100.0
CAS Min % 96.0
Molecular Formula C11H17NO4
Assay Percent Range 97%
Specific Rotation +240° to +260° (20°C, 589nm) c=0.5, methanol
Chemical Name or Material (1R, 4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid
CAS 298716-03-7
Infrared Spectrum Authentic