Please login to your online account to display your discounted pricing

(1R,4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC435880050

 View more versions of this product

Catalog No. AC435880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 227.26
Quantity 5g
Color White
Melting Point 110° to 115°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (1R, 4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid
Specific Rotation +240° to +260° (20°C, 589nm) c=0.5, methanol
Molecular Formula C11H17NO4
CAS 298716-03-7
Infrared Spectrum Authentic
Assay Percent Range 97%

Specifications

Chemical Name or Material (1R, 4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid
CAS 298716-03-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H17NO4
Formula Weight 227.26