Please login to your online account to display your discounted pricing

(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% e.e., ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361832500

 View more versions of this product

Catalog No. AC361832500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 229.27
Physical Form Powder or Chunks
Quantity 250mg
Color White
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Synonym (1R, 3S)-N-tert-Butoxycarbonyl-1-aminocyclopentane-3-carboxylic acid
Infrared Spectrum Authentic
MDL Number MFCD01320857
Molecular Formula C11H19NO4
Assay Percent Range 95%
CAS 261165-05-3
Chemical Name or Material (1R, 3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid
Name Note 95%, (98% e.e.)
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)

Specifications

Chemical Name or Material (1R, 3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid
Name Note 95%, (98% e.e.)
CAS 261165-05-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H19NO4
Formula Weight 229.27
MDL Number MFCD01320857
Synonym (1R, 3S)-N-tert-Butoxycarbonyl-1-aminocyclopentane-3-carboxylic acid