Please login to your online account to display your discounted pricing

(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilzed, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399342500

 View more versions of this product

Catalog No. AC399342500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid
Assay ≥98.5% (GC)
Formula Weight 142.15
Quantity 250mg
Color Off-white to beige
Melting Point 49° to 53°C
CAS Max % 100.0
Chemical Name or Material (1R, 3S)-(+)-4-Cyclopentene-1, 3-diol 1-acetate
Packaging Glass bottle
Name Note stabilzed
Assay Percent Range 99%
Molecular Formula C7H10O3
Specific Rotation +67° to +69° (20°C, 589nm) ((c=2.3 in chloroform))
MDL Number MFCD00078933
CAS 60410-16-4
CAS Min % 98.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (1R, 3S)-(+)-4-Cyclopentene-1, 3-diol 1-acetate
Name Note stabilzed
CAS 60410-16-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C7H10O3
Formula Weight 142.15
MDL Number MFCD00078933