missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1R,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 362395000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(1R, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid | |
489446-85-7 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD02259727 | |
1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid | |
RNJQBGXOSAQQDG-HTQZYQBOSA-N | |
(1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid | |
1512527 | |
95%,98% ee |
95%, (98% e.e.) | |
94.0 | |
97.5% min. | |
94% min. (GC) | |
C11H19NO4 | |
500mg | |
Solubility in water: soluble | |
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O | |
229.27 | |
229.27 |
Chemical Identifiers
489446-85-7 | |
229.27 | |
RNJQBGXOSAQQDG-HTQZYQBOSA-N | |
1512527 | |
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O |
C11H19NO4 | |
MFCD02259727 | |
1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid | |
(1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid |
Safety and Handling
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
TSCA : TSCA
RUO â Research Use Only