missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 362390010

 View more versions of this product

Catalog No. AC362390010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(1R, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid
Authentic
White to Yellow
1g
489446-85-7
100.0
C11H19NO4
1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid
RNJQBGXOSAQQDG-HTQZYQBOSA-N
(1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
1512527
Powder
97.5% min.
Glass bottle
117.3°C
95%, (98% e.e.)
94.0
94% min. (GC)
MFCD02259727
Solubility in water: soluble
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O
229.27
229.27
95%,98% ee

Chemical Identifiers

489446-85-7
229.27
RNJQBGXOSAQQDG-HTQZYQBOSA-N
1512527
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O
C11H19NO4
MFCD02259727
1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid
(1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
Safety and Handling

Safety and Handling

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

TSCA : TSCA

SDS Documents