Please login to your online account to display your discounted pricing

(1R,2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol, 99%, 99% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369490010

 View more versions of this product

Catalog No. AC369490010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 219.33
Physical Form Crystalline Powder
Assay ≥98.5%
Quantity 1g
Color White to off-white
Melting Point 66° to 70°C
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula C6H5CHNHC6H10CHOH
Name Note 99% e.e.
CAS 71581-92-5
Molecular Formula C14H21NO
Chemical Name or Material (1R, 2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol
Assay Percent Range 99%
MDL Number MFCD00151395
CAS Min % 98.5
Packaging Glass bottle
Enantiomeric Excess 98.5% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material (1R, 2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol
Name Note 99% e.e.
CAS 71581-92-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14H21NO
Linear Formula C6H5CHNHC6H10CHOH
Formula Weight 219.33
MDL Number MFCD00151395
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents