Please login to your online account to display your discounted pricing

(1R,2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol, 99%, 99% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1R, 2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol
Name Note 99% e.e.
CAS 71581-92-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14H21NO
Linear Formula C6H5CHNHC6H10CHOH
Formula Weight 219.33
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369490010

  SDS
thermo fisher scientific
AC369490010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC369490050

  SDS
thermo fisher scientific
AC369490050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.33
Physical Form Crystalline Powder
Assay ≥98.5%
Quantity 1g
Color White to off-white
Melting Point 66° to 70°C
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula C6H5CHNHC6H10CHOH
Name Note 99% e.e.
CAS 71581-92-5
Molecular Formula C14H21NO
Chemical Name or Material (1R, 2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol
Assay Percent Range 99%
MDL Number MFCD00151395
CAS Min % 98.5
Packaging Glass bottle
Enantiomeric Excess 98.5% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.