missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate, TCI America™

$149.72 - $508.77

Chemical Identifiers

CAS 1517-82-4
Molecular Formula C17H26O2S
Molecular Weight (g/mol) 294.453
MDL Number MFCD00010192
InChI Key NQICGNSARVCSGJ-DSRNLFJRSA-N
Synonym 1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 101348873
IUPAC Name [(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
SMILES CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10445G View Documents TCI America
M10445G
5g Each for $149.72
 
M104425G View Documents TCI America
M104425G
25g Each for $508.77
 
Specifications

Chemical Identifiers

1517-82-4
294.453
NQICGNSARVCSGJ-DSRNLFJRSA-N
101348873
CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C
C17H26O2S
MFCD00010192
1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
[(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate

Specifications

White-Yellow
5g
C17H26O2S
1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C
294.453
294.45
≥98.0% (HPLC)
105°C
1517-82-4
MFCD00010192
NQICGNSARVCSGJ-DSRNLFJRSA-N
[(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
101348873
Crystal-Powder at 20°C
(1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer