missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl (S)-2-Hydroxypropionate, TCI America™

$55.00 - $172.64

Chemical Identifiers

CAS 61597-98-6
Molecular Formula C13H24O3
Molecular Weight (g/mol) 228.332
InChI Key UJNOLBSYLSYIBM-NOOOWODRSA-N
Synonym (S)-2-Hydroxypropionic Acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, L-Menthyl L-Lactate
PubChem CID 7076215
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2S)-2-hydroxypropanoate
SMILES CC1CCC(C(C1)OC(=O)C(C)O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08895G View Documents TCI America
I08895G
5g Each for $55.00
 
I088925G View Documents TCI America
I088925G
25g Each for $172.64
 
Specifications

Chemical Identifiers

61597-98-6
228.332
(S)-2-Hydroxypropionic Acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, L-Menthyl L-Lactate
[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2S)-2-hydroxypropanoate
C13H24O3
UJNOLBSYLSYIBM-NOOOWODRSA-N
7076215
CC1CCC(C(C1)OC(=O)C(C)O)C(C)C

Specifications

White
5g
C13H24O3
UJNOLBSYLSYIBM-NOOOWODRSA-N
[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2S)-2-hydroxypropanoate
7076215
Crystal-Powder at 20°C
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl (S)-2-Hydroxypropionate
45°C
61597-98-6
(S)-2-Hydroxypropionic Acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, L-Menthyl L-Lactate
CC1CCC(C(C1)OC(=O)C(C)O)C(C)C
228.332
228.33
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3279

TSCA : No