Please login to your online account to display your discounted pricing

(1R,2S,4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass Bottle in Plastic Container

Manufacturer: thermo fisher scientific  362295000

 View more versions of this product

Catalog No. AC362295000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 259.3
Quantity 500mg
Color Clear colorless to yellow
Packaging Glass Bottle in Plastic Container
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Chemical Name or Material (1R, 2S, 4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
Molecular Formula C12H21NO5
MDL Number MFCD02259718
CAS 321744-14-3
Synonym Methyl (1S, 3R, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (1R, 2S, 4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
CAS 321744-14-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD02259718
Synonym Methyl (1S, 3R, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate