Please login to your online account to display your discounted pricing

(1R,2S,4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1R, 2S, 4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
CAS 321744-14-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD02259718
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362295000

  SDS
thermo fisher scientific
362295000
Glass Bottle in Plastic Container 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 259.3
Physical Form Viscous Liquid
Quantity 500mg
Color Clear colorless to yellow
Packaging Glass Bottle in Plastic Container
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Assay Percent Range 95%
CAS 321744-14-3
Name Note 95%, (98% e.e.)
Molecular Formula C12H21NO5
Chemical Name or Material (1R, 2S, 4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
MDL Number MFCD02259718
Enantiomeric Excess 97.5% min. (GC)
Synonym Methyl (1S, 3R, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate
Infrared Spectrum Authentic