Please login to your online account to display your discounted pricing

(1R,2S,4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic Acid Methyl Ester 95% (98% ee), Acros Organics


Quantity 500mg

Manufacturer: thermo fisher scientific  AC362335000

 View more versions of this product

Catalog No. AC362335000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 259.3
Physical Form Crystalline Powder
Quantity 500mg
Color Yellow
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Chemical Name or Material (1R, 2S, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
CAS 321744-17-6
Name Note 95%, (98% e.e.)
Molecular Formula C12H21NO5
MDL Number MFCD02259722
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Synonym Methyl (1R, 3R, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate

Specifications

Chemical Name or Material (1R, 2S, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester
Name Note 95%, (98% e.e.)
CAS 321744-17-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD02259722
Synonym Methyl (1R, 3R, 4S)-N-BOC-3-amino-4-hydroxycyclopentanecarboxylate