Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2S)-(-)-2,N-Epoxy-10,2-camphorsultam, 96%

Chemical Identifiers

Chemical Name or Material (1R,2S)-(-)-2,N-Epoxy-10,2-camphorsultam
CAS 104372-31-8
Formula Weight 229.3
MDL Number MFCD00075428
Beilstein 6274369
Synonym (1R)-(-)-(10-Camphorsulfonyl)oxaziridine; (1R)-(-)-2,N-Epoxy-exo-10,2-bornanesultam
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5954403

  SDS
alfa aesar™
H5954403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.3
MDL Number MFCD00075428
Color White
Melting Point 170°C
Chemical Name or Material (1R,2S)-(-)-2,N-Epoxy-10,2-camphorsultam
CAS 104372-31-8
Beilstein 6274369
Physical Form Powder
Synonym (1R)-(-)-(10-Camphorsulfonyl)oxaziridine; (1R)-(-)-2,N-Epoxy-exo-10,2-bornanesultam
Assay 96%
Storage Note 1 -30° to -10°C
Quantity 1g
Safety and Handling

-30° to -10°C