missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol, TCI America™

$186.39 - $630.98

Chemical Identifiers

CAS 187324-63-6
Molecular Formula C25H29NO3S
Molecular Weight (g/mol) 423.571
MDL Number MFCD02093446
InChI Key ZPERKXJNXPLVAC-URXFXBBRSA-N
Synonym (1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol
PubChem CID 11166154
IUPAC Name N-benzyl-N-[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]-2,4,6-trimethylbenzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)N(CC2=CC=CC=C2)C(C)C(C3=CC=CC=C3)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21035G View Documents TCI America
B21035G
5g Each for $630.98
 
B21031G View Documents TCI America
B21031G
1g Each for $186.39
 
Specifications

Chemical Identifiers

187324-63-6
423.571
ZPERKXJNXPLVAC-URXFXBBRSA-N
11166154
CC1=CC(=C(C(=C1)C)S(=O)(=O)N(CC2=CC=CC=C2)C(C)C(C3=CC=CC=C3)O)C
C25H29NO3S
MFCD02093446
(1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol
N-benzyl-N-[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]-2,4,6-trimethylbenzenesulfonamide

Specifications

White-Yellow
5g
C25H29NO3S
(1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol
CC1=CC(=C(C(=C1)C)S(=O)(=O)N(CC2=CC=CC=C2)C(C)C(C3=CC=CC=C3)O)C
423.571
423.57
≥98.0% (HPLC,N)
132°C
187324-63-6
MFCD02093446
ZPERKXJNXPLVAC-URXFXBBRSA-N
N-benzyl-N-[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]-2,4,6-trimethylbenzenesulfonamide
11166154
Crystal-Powder at 20°C
(1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol
Safety and Handling

Safety and Handling

TSCA : No