Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2S)-(-)-2-Amino-1,2-diphenylethanol, 99%

$74.65 - $263.22

Chemical Identifiers

Chemical Name or Material (1R,2S)-(-)-2-Amino-1,2-diphenylethanol
CAS 23190-16-1
Molecular Formula C14H15NO
Formula Weight 213.28
MDL Number MFCD00074960
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2760703

 
alfa aesar™
H2760703
1g Each for $74.65

AAH2760706

 
alfa aesar™
H2760706
5g Each for $263.22
Description & Specifications

Specifications

Formula Weight 213.28
Chemical Name or Material (1R,2S)-(-)-2-Amino-1,2-diphenylethanol
Assay 99%
Molecular Formula C14H15NO
MDL Number MFCD00074960
CAS 23190-16-1
Melting Point 141° to 143°C
Quantity 1g
Optical Rotation −6.9° (c=0.6, Ethanol)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.