Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-Bis(4-methoxyphenyl)-1,2-ethanediamine, 98%

$98.24 - $1,069.80

Chemical Identifiers

Chemical Name or Material (1R,2R)-Bis(4-methoxyphenyl)-1,2-ethanediamine
CAS 58520-03-9
Molecular Formula C16H20N2O2
Formula Weight 272.34
MDL Number MFCD08460192
Synonym (1R,2R)-Bis(4-methoxyphenyl)-1,2-diaminoethane; (R,R)-DAEN
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3108103

  SDS
alfa aesar™
H3108103
1g Each for $98.24

AAH3108106

  SDS
alfa aesar™
H3108106
5g Each for $322.45

AAH3108114

  SDS
alfa aesar™
H3108114
25g Each for $1,069.80
Description & Specifications

Specifications

Formula Weight 272.34
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (1R,2R)-Bis(4-methoxyphenyl)-1,2-ethanediamine
Molecular Formula C16H20N2O2
MDL Number MFCD08460192
CAS 58520-03-9
Melting Point 89° to 91°C
Health Hazard 1 Warning
Optical Rotation +77° (c=2 in Chloroform)
Synonym (1R,2R)-Bis(4-methoxyphenyl)-1,2-diaminoethane; (R,R)-DAEN
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.