missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1R,2R,3S,5R)-(-)-2,3-Pinanediol, 99%, ACROS Organics™
Chemical Identifiers
CAS | 22422-34-0 |
---|---|
Molecular Formula | C10H18O2 |
Molecular Weight (g/mol) | 170.25 |
InChI Key | MOILFCKRQFQVFS-BDNRQGISSA-N |
Synonym | --2,3-pinanediol |
PubChem CID | 6553875 |
IUPAC Name | (1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol |
SMILES | CC1(C2CC1C(C(C2)O)(C)O)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC279560010 SDS |
Thermo Fisher Scientific
AC279560010 |
1g | Glass bottle | N/A | N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
22422-34-0 | |
170.25 | |
--2,3-pinanediol | |
(1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol |
C10H18O2 | |
MOILFCKRQFQVFS-BDNRQGISSA-N | |
6553875 | |
CC1(C2CC1C(C(C2)O)(C)O)C |
Specifications
>110°C | |
Glass bottle | |
− 8.50 | |
Colorless to Yellow | |
1g | |
98.5 | |
98.5% min. (GC) | |
06,4145 | |
MOILFCKRQFQVFS-BDNRQGISSA-N | |
(1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol | |
6553875 | |
Liquid | |
(1R,2R,3S,5R)-(-)-2,3-Pinanediol |
Authentic | |
−8.50 (20.00°C c=6.5,toluene) | |
101.0°C to 102.0°C (1.0 mmHg) | |
55.0°C to 60.0°C | |
22422-34-0 | |
100.0 | |
C10H18O2 | |
--2,3-pinanediol | |
CC1(C2CC1C(C(C2)O)(C)O)C | |
170.25 | |
170.25 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning