missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1R,2R,3S,5R)-(-)-2,3-Pinanediol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC279560010
Specifications
(1R,2R,3S,5R)-(-)-2,3-Pinanediol | |
Authentic | |
−8.50 (20.00°C c=6.5,toluene) | |
101.0°C to 102.0°C (1.0 mmHg) | |
55.0°C to 60.0°C | |
22422-34-0 | |
100.0 | |
C10H18O2 | |
--2,3-pinanediol, --2-hydroxyisopinocampheol, --cis-pinane-cis-2,3-diol, 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r, pinanediol, unii-r58l0w3a75 | |
CC1(C2CC1C(C(C2)O)(C)O)C | |
170.25 | |
170.25 | |
99% |
>110°C | |
Glass bottle | |
− 8.50 | |
Colorless to Yellow | |
1g | |
98.5 | |
98.5% min. (GC) | |
06,4145 | |
MOILFCKRQFQVFS-BDNRQGISSA-N | |
(1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol | |
6553875 | |
Liquid |
Chemical Identifiers
22422-34-0 | |
170.25 | |
--2,3-pinanediol, --2-hydroxyisopinocampheol, --cis-pinane-cis-2,3-diol, 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r, pinanediol, unii-r58l0w3a75 | |
(1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol |
C10H18O2 | |
MOILFCKRQFQVFS-BDNRQGISSA-N | |
6553875 | |
CC1(C2CC1C(C(C2)O)(C)O)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning