missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439540010

 View more versions of this product

Catalog No. AC439540010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(1R,2R,3R,5S)-(-)-Isopinocampheylamine
Authentic
-42
Colorless to Yellow
69460-11-3
100.0
C10H19N
VPTSZLVPZCTAHZ-BZNPZCIMSA-N
(1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
11744832
Liquid
72°C
Glass bottle
90°C (18.0mmHg)
1g
94.0
94% min. (GC)
--isopinocampheyl amine, --isopinocampheylamine, --isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine, 1r,2r,3r,5s---isopinocampheylamine, 1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine, 1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine, 1r,2r,3r,5s-3-pinanamine, 1r,2r,5s-pinane-3alpha-amine, bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s
CC1C2CC(C2(C)C)CC1N
153.27
153.27
95%

Chemical Identifiers

69460-11-3
153.27
--isopinocampheyl amine, --isopinocampheylamine, --isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine, 1r,2r,3r,5s---isopinocampheylamine, 1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine, 1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine, 1r,2r,3r,5s-3-pinanamine, 1r,2r,5s-pinane-3alpha-amine, bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s
(1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
C10H19N
VPTSZLVPZCTAHZ-BZNPZCIMSA-N
11744832
CC1C2CC(C2(C)C)CC1N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS Documents