Please login to your online account to display your discounted pricing

(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (1R,2R,3R,5S)-(-)-Isopinocampheylamine
CAS 69460-11-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C10H19N
Formula Weight 153.27
Synonym (1R, 2R, 3R
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC439540050

 
thermo fisher scientific
AC439540050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439540010

 
thermo fisher scientific
AC439540010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥94.0%
Formula Weight 153.27
Quantity 5g
Color Colorless to yellow
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation.
Specific Rotation -38.0° to -44.0° (20°C, 589nm) (Neat)
Chemical Name or Material (1R,2R,3R,5S)-(-)-Isopinocampheylamine
Assay Percent Range 95%
CAS 69460-11-3
Molecular Formula C10H19N
Synonym (1R, 2R, 3R
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le
Safety and Handling

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le

GHS P Statement Causes skin irritation. Causes serious eye irritation.