Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(-)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 98%

$229.35 - $229.35

Chemical Identifiers

Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclopentylamine
CAS 181657-56-7
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD01075750
UN Number UN2735
Synonym (1R,2R)-1-Amino-2-benzyloxycyclopentane; (1R-trans)-2-(Phenylmethoxy)cyclopentanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1701706

 
alfa aesar™
L1701706
5g Each for $229.35

AAL1701703

 
alfa aesar™
L1701703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.27
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclopentylamine
Assay ≥99%
Molecular Formula C12H17NO
MDL Number MFCD01075750
CAS 181657-56-7
Boiling Point 90° to 93°C (0.23mmHg)
Melting Point 23°C
Synonym (1R,2R)-1-Amino-2-benzyloxycyclopentane; (1R-trans)-2-(Phenylmethoxy)cyclopentanamine
Quantity 5g
Flash Point >110°C (230°F)
Optical Rotation −57.5° (Neat)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Assay Percent Notes ee 98%
UN Number UN2735

Applications
(1R,2R)-(-)-2-Benzyloxycyclopentylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Not miscible or difficult to mix with water.

Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.