Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(-)-2-Benzyloxycyclopentyl isocyanate, 96%

Chemical Identifiers

Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclopentyl isocyanate
CAS 737001-14-8
Molecular Formula C13H15NO2
Formula Weight 217.27
MDL Number MFCD05664069
UN Number UN2206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2022403

 
alfa aesar™
L2022403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.27
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclopentyl isocyanate
Molecular Formula C13H15NO2
CAS 737001-14-8
Assay 96%
MDL Number MFCD05664069
UN Number UN2206
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C