Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(-)-2-Benzyloxycyclohexylamine, ChiPros 98+%, ee 98%

$224.51 - $224.51

Chemical Identifiers

Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexylamine
CAS 216394-06-8
Molecular Formula C13H19NO
Formula Weight 205.3
MDL Number MFCD01075752
UN Number UN2735
Synonym (1R,2R)-(-)-1-Amino-2-benzyloxycyclohexane; (1R-trans)-(-)-2-(Phenylmethoxy)cyclohexanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1701906

  SDS
alfa aesar™
L1701906
5g Each for $224.51

AAL1701903

  SDS
alfa aesar™
L1701903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.3
Health Hazard 1 Danger
CAS 216394-06-8
UN Number UN2735
Boiling Point 79° to 80°C (0.08mmHg)
Optical Rotation −85° (Neat)
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym (1R,2R)-(-)-1-Amino-2-benzyloxycyclohexane; (1R-trans)-(-)-2-(Phenylmethoxy)cyclohexanamine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexylamine
Molecular Formula C13H19NO
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Quantity 5g
Recommended Storage 2° to 8°C
Assay ≥98%
Assay Percent Notes ee 98%
MDL Number MFCD01075752
Flash Point >110°C (230°F)

Applications
(1R,2R)-(-)-2-Benzyloxycyclohexylamine is used as an pharma intermediates.

Solubility
Difficult to mix.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C