Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(-)-2-Benzyloxycyclohexyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexyl isothiocyanate
CAS 745783-98-6
Molecular Formula C14H17NOS
Formula Weight 247.36
MDL Number MFCD05664116
UN Number UN2923
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2031603

 
alfa aesar™
L2031603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.36
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexyl isothiocyanate
Assay 97%
Molecular Formula C14H17NOS
MDL Number MFCD05664116
CAS 745783-98-6
UN Number UN2923
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Melting Point 33° to 36°C
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

2° to 8°C