Please login to your online account to display your discounted pricing

Alfa Aesar™ (1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate
CAS 745784-13-8
Molecular Formula C14H17NO
Formula Weight 231.3
MDL Number MFCD05664133
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2035303

  SDS
alfa aesar™
L2035303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.3
MDL Number MFCD05664133
Health Hazard 2 GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Danger
Melting Point 35° to 37°C
Molecular Formula C14H17NO
Assay 97%
Quantity 1g
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Recommended Storage 2° to 8°C
CAS 745784-13-8
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C