missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, ACROS Organics™

$64.85 - $178.90

Chemical Identifiers

CAS 716-61-0
Molecular Formula C9H13N2O4
Molecular Weight (g/mol) 213.21
MDL Number MFCD00078126
InChI Key OCYJXSUPZMNXEN-RKDXNWHRSA-O
Synonym 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r, 1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--, 1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol, 1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol, 1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol, chloramphenicol base, d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol, d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol, d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol, levoamine
PubChem CID 69745
ChEBI CHEBI:78660
IUPAC Name (1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
SMILES [NH3+][C@H](CO)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334790050
SDS View Documents
Thermo Fisher Scientific
334790050
5g Glass bottle
Each for $64.85
 
AC334790250
SDS View Documents
Thermo Fisher Scientific
334790250
25g Glass bottle
Each for $178.90
 
Description

Description

Specifications

Chemical Identifiers

716-61-0
213.21
OCYJXSUPZMNXEN-RKDXNWHRSA-O
69745
(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
C9H13N2O4
MFCD00078126
1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r, 1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--, 1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol, 1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol, 1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol, chloramphenicol base, d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol, d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol, d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol, levoamine
CHEBI:78660
[NH3+][C@H](CO)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Authentic
-31
160°C to 166°C
716-61-0
100.0
C9H13N2O4
1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r, 1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--, 1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol, 1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol, 1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol, chloramphenicol base, d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol, d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol, d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol, levoamine
[NH3+][C@H](CO)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O
213.21
CHEBI:78660
Powder
(1R, 2R)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol, 96%
Glass bottle
Yellow
5g
95.0
96%
MFCD00078126
OCYJXSUPZMNXEN-RKDXNWHRSA-O
(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
69745
212.2
96%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 211-938-7

RTECSNumber : TY3100000

TSCA : TSCA

SDS
Product Certifications