missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2R)-2-[(4S,11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]-1,2-diphenylethanammonium tetrafluoroborate, 97+%, Alfa Aesar™

Chemical Identifiers

Molecular Formula C36H31BF4NP
MDL Number MFCD17018775
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60082MD View Documents Alfa Aesar
H60082MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6008203 View Documents Alfa Aesar
H6008203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

C36H31BF4NP
MFCD17018775

Specifications

250mg
MFCD17018775
≥97%
C36H31BF4NP
595.42
(1R,2R)-2-[(4S,11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]-1,2-diphenylethanammonium tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning